Assay Details
| Gene Symbol: Apoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013475.4 | NP_038503.4 | 2 - 3 | 289 | 130 | |
| GenBank mRNA | AK010926.1 | - | 2 - 3 | 277 | 130 | |
| BC011037.1 | - | 2 - 3 | 305 | 130 | ||
| BC019811.1 | - | 2 - 3 | 277 | 5125770 | 130 | |
| BC053338.1 | - | 2 - 3 | 265 | 5125963 | 130 | |
| D10056.1 | - | 2 - 3 | 263 | 130 | ||
| S70439.1 | - | 2 - 3 | 273 | 130 | ||
More Information
Set Membership: |
|

Apoh
Genomic Map