| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100017070.mm.1 |
NM_013475.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017070.mm.1 |
NM_013475.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017070.mm.1 |
TC1100001813.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017072.mm.1 |
NM_013475.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017072.mm.1 |
NM_013475.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017072.mm.1 |
TC1100001813.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017074.mm.1 |
NM_013475.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017074.mm.1 |
NM_013475.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017074.mm.1 |
TC1100001813.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017071.mm.1 |
NM_013475.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017071.mm.1 |
NM_013475.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017071.mm.1 |
TC1100001813.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017075.mm.1 |
NM_013475.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017075.mm.1 |
NM_013475.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017075.mm.1 |
TC1100001813.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100017073.mm.1 |
NM_013475.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017073.mm.1 |
NM_013475.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100017073.mm.1 |
TC1100001813.mm.1 |
MTA TC |