| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100040595.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040595.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040595.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040599.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040599.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040599.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040600.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040600.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040600.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040605.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040605.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040605.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040612.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040612.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040612.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040592.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040592.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040592.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040608.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040608.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040608.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040590.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040590.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040590.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040604.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040604.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040604.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040606.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040606.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040606.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040589.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040589.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040589.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040601.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040601.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040601.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040610.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040610.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040610.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040602.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040602.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040602.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040593.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040593.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040593.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040596.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040596.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040596.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040603.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040603.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040603.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040607.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040607.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040607.mm.1 |
TC1100004234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100040609.mm.1 |
NM_027745.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040609.mm.1 |
NM_027745.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100040609.mm.1 |
TC1100004234.mm.1 |
MTA TC |