Assay Details
| Gene Symbol: Ccdc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027745.1 | NP_082021.1 | 17 - 18 | 2782 | 120 | |
| GenBank mRNA | AK017054.1 | - | 2 - 3 | 249 | 120 | |
| BC030385.1 | - | 4 - 5 | 569 | 120 | ||
| BC056480.1 | - | 17 - 18 | 2782 | 5689542 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017054.1 transcripts.
Set Membership: |
|

Ccdc57
Genomic Map