Assay Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029746.3 | NP_084022.2 | 6 - 7 | 708 | 70 | |
| GenBank mRNA | AK089920.1 | - | 6 - 7 | 692 | 70 | |
| AK157348.1 | - | 6 - 7 | 670 | 70 | ||
| AK169298.1 | - | 6 - 7 | 678 | 70 | ||
| AK171726.1 | - | 6 - 7 | 627 | 70 | ||
| BC011435.1 | - | 6 - 7 | 648 | 4010102 | 70 | |
| CO423795.1 | - | 3 - 4 | 265 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cog2
Genomic Map