Assay Details
| Gene Symbol: Psmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010817.2 | NP_034947.1 | 2 - 3 | 295 | 145 | |
| GenBank mRNA | AK031013.1 | - | 2 - 3 | 246 | 145 | |
| AK159825.1 | - | 2 - 3 | 247 | 145 | ||
| AK217500.1 | - | 1 - 2 | 74 | 145 | ||
| BC013513.1 | - | 2 - 3 | 277 | 145 | ||
| BC130014.1 | - | 2 - 3 | 272 | 40140405 | 145 | |
| M64641.1 | - | 2 - 3 | 243 | 145 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmd7
Genomic Map