Assay Details
| Gene Symbol: Nob1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026277.3 | NP_080553.1 | 6 - 7 | 705 | 64 | |
| GenBank mRNA | AK021194.1 | - | 6 - 7 | 709 | 64 | |
| AK075755.1 | - | 6 - 7 | 706 | 64 | ||
| AK211187.1 | - | 1 - 2 | 36 | 64 | ||
| BC034900.1 | - | 6 - 7 | 705 | 64 | ||
| BC041150.1 | - | 4 - 5 | 466 | 64 | ||
| BC054835.1 | - | 6 - 7 | 659 | 64 | ||
| BC103793.1 | - | 6 - 7 | 708 | 6328184 | 64 | |
| BC117992.2 | - | 6 - 7 | 706 | 40104531 | 64 | |
| BC117993.2 | - | 6 - 7 | 706 | 40104533 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nob1
Genomic Map