Assay Details
| Gene Symbol: Ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025660.2 | NP_079936.1 | 2 - 3 | 229 | 72 | |
| GenBank mRNA | AK011592.1 | - | 2 - 3 | 229 | 72 | |
| AK017461.1 | - | 2 - 3 | 121 | 72 | ||
| BC050755.1 | - | 2 - 3 | 217 | 6772058 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011592.1, AK017461.1, BC050755.1, NM_025660.2 transcripts.
Set Membership: |
|

Ribc1
Genomic Map