Assay Details
| Gene Symbol: B430212C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033214.1 | 2 - 3 | 337 | 71 | ||
| GenBank mRNA | AK087569.1 | - | 2 - 3 | 338 | 71 | |
| AK148585.1 | - | 2 - 3 | 496 | 71 | ||
| AK164742.1 | - | 2 - 3 | 304 | 71 | ||
| BC028804.1 | - | 2 - 3 | 245 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | inositol (myo)-1(or 4)-monophosphatase 2 | Impa2 |
2210415D20Rik, AI326924, AW259601 |
-5.0kb | Anti-sense | Chr. 18 - 67289223 - 67318841 on NCBI Build GRCm38 |
| Downstream |
cell death-inducing DNA fragmentation factor, alpha subunit-like effector A |
Cidea | AW212747 | 18.8kb | Anti-sense | Chr. 18 - 67343564 - 67367794 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

B430212C06Rik
Genomic Map