| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118473 |
AK046629.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118471 |
AK164742.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118471 |
AK148585.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118471 |
AK046629.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118471 |
AK087569.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2118471 |
NR_033214.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895444 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836615 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
650646 |
AK089133.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
650646 |
AK163637.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
148565 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914589 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
452426 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
630596 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
19909 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
662741 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325294 |
AK131902.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325294 |
AK133712.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325294 |
AK140957.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325294 |
BC048089.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325294 |
NM_001012667.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
419967 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
287457 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
868840 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
41621 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
103084 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
4669 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P110806 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P116336 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118471 |
AK164742.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118471 |
AK148585.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118471 |
AK046629.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118471 |
AK087569.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118471 |
NR_033214.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2118473 |
AK046629.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P110806 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09036261 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09036275 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P116336 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P116336 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P110806 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791304 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13803351 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791294 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14078085 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791290 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13855881 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791313 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791302 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791292 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791299 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13853325 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13855884 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791301 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31791307 |
- |
- |