Assay Details
| Gene Symbol: Tubb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026473.2 | NP_080749.2 | 2 - 3 | 188 | 62 | |
| GenBank mRNA | AK009958.1 | - | 2 - 3 | 199 | 62 | |
| AK144815.1 | - | 2 - 3 | 203 | 62 | ||
| BC008225.1 | - | 2 - 3 | 188 | 2631779 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tubb6
Genomic Map