Assay Details
| Gene Symbol: Gtf3c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033194.2 | NP_001028366.1 | 11 - 12 | 1642 | 88 | |
| GenBank mRNA | AK145040.1 | - | 1644 | 89 | ||
| AK160997.1 | - | 11 - 12 | 1630 | 88 | ||
| AK165830.1 | - | 11 - 12 | 1643 | 88 | ||
| BC141157.1 | - | 11 - 12 | 1632 | 9055769 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gtf3c3
Genomic Map