Assay Details
| Gene Symbol: Gtf3c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033194.2 | NP_001028366.1 | 16 - 17 | 2429 | 96 | |
| GenBank mRNA | AK145040.1 | - | 2433 | 96 | ||
| AK160997.1 | - | 16 - 17 | 2417 | 96 | ||
| AK165830.1 | - | 16 - 17 | 2430 | 96 | ||
| BC141157.1 | - | 16 - 17 | 2419 | 9055769 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gtf3c3
Genomic Map