Assay Details
| Gene Symbol: Gtf3c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033194.2 | NP_001028366.1 | 8 - 9 | 1162 | 64 | |
| GenBank mRNA | AK145040.1 | - | 1164 | 64 | ||
| AK160997.1 | - | 8 - 9 | 1150 | 64 | ||
| AK165830.1 | - | 8 - 9 | 1163 | 64 | ||
| AK205225.1 | - | 1 - 2 | 86 | 64 | ||
| BC141157.1 | - | 8 - 9 | 1152 | 9055769 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gtf3c3
Genomic Map