Assay Details
| Gene Symbol: Mdh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029696.4 | NP_083972.3 | 3 - 4 | 293 | 68 | |
| GenBank mRNA | BC050786.1 | - | 3 - 4 | 293 | 6703279 | 68 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC050786.1 transcripts.
Set Membership: |
|

Mdh1b
Genomic Map