Assay Details
| Gene Symbol: Tmbim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027154.5 | NP_081430.3 | 4 - 5 | 618 | 66 | |
| GenBank mRNA | AB097685.1 | - | 3 - 4 | 364 | 66 | |
| AK077689.1 | - | 4 - 5 | 443 | 66 | ||
| AK144110.1 | - | 4 - 5 | 462 | 66 | ||
| AK147297.1 | - | 3 - 4 | 9326 | 66 | ||
| AK152864.1 | - | 4 - 5 | 461 | 66 | ||
| AK166926.1 | - | 4 - 5 | 634 | 66 | ||
| AK170625.1 | - | 4 - 5 | 428 | 66 | ||
| AK171102.1 | - | 4 - 5 | 460 | 66 | ||
| AK220296.1 | - | 4 - 5 | 575 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmbim1
Genomic Map