Assay Details
| Gene Symbol: Tmbim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027154.5 | NP_081430.3 | 7 - 8 | 763 | 76 | |
| GenBank mRNA | AB097685.1 | - | 6 - 7 | 509 | 76 | |
| AK144110.1 | - | 7 - 8 | 607 | 76 | ||
| AK147297.1 | - | 6 - 7 | 9471 | 76 | ||
| AK152864.1 | - | 7 - 8 | 606 | 76 | ||
| AK167286.1 | - | 2 - 3 | 62 | 76 | ||
| AK170625.1 | - | 7 - 8 | 573 | 76 | ||
| AK171102.1 | - | 7 - 8 | 605 | 76 | ||
| AK180422.1 | - | 2 - 3 | 102 | 76 | ||
| AK186797.1 | - | 2 - 3 | 119 | 76 | ||
| AK188714.1 | - | 2 - 3 | 106 | 76 | ||
| AK220296.1 | - | 5 - 6 | 1769 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmbim1
Genomic Map