Assay Details
| Gene Symbol: Tmbim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027154.5 | NP_081430.3 | 8 - 9 | 791 | 71 | |
| GenBank mRNA | AB097685.1 | - | 7 - 8 | 537 | 71 | |
| AK144110.1 | - | 8 - 9 | 635 | 71 | ||
| AK147297.1 | - | 7 - 8 | 9499 | 71 | ||
| AK152864.1 | - | 8 - 9 | 634 | 71 | ||
| AK167286.1 | - | 3 - 4 | 90 | 71 | ||
| AK170625.1 | - | 8 - 9 | 601 | 71 | ||
| AK171102.1 | - | 8 - 9 | 633 | 71 | ||
| AK188714.1 | - | 3 - 4 | 134 | 71 | ||
| AK220296.1 | - | 6 - 7 | 1797 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmbim1
Genomic Map