Assay Details
| Gene Symbol: Ctdsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153088.2 | NP_694728.1 | 3 - 4 | 614 | 91 | |
| GenBank mRNA | AK089478.1 | - | 3 - 4 | 522 | 91 | |
| AK167390.1 | - | 3 - 4 | 372 | 91 | ||
| AY028804.1 | - | 3 - 4 | 639 | 91 | ||
| BC049184.1 | - | 3 - 4 | 474 | 6439402 | 91 | |
| BC058085.1 | - | 2 - 3 | 801 | 91 | ||
| BC065158.1 | - | 3 - 4 | 349 | 5717102 | 91 | |
| BC079638.1 | - | 3 - 4 | 349 | 5717102 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctdsp1
Genomic Map