Assay Details
| Gene Symbol: Ctdsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153088.2 | NP_694728.1 | 4 - 5 | 666 | 88 | |
| GenBank mRNA | AK089478.1 | - | 4 - 5 | 574 | 88 | |
| AK167390.1 | - | 4 - 5 | 424 | 88 | ||
| AY028804.1 | - | 4 - 5 | 691 | 88 | ||
| BC049184.1 | - | 4 - 5 | 526 | 6439402 | 88 | |
| BC058085.1 | - | 3 - 4 | 853 | 88 | ||
| BC065158.1 | - | 4 - 5 | 401 | 5717102 | 88 | |
| BC079638.1 | - | 4 - 5 | 401 | 5717102 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctdsp1
Genomic Map