Assay Details
| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160038.1 | NP_001153510.1 | 16 - 17 | 2212 | 65 | |
| NM_001160039.1 | NP_001153511.1 | 16 - 17 | 2054 | 65 | ||
| NM_001160040.1 | NP_001153512.1 | 15 - 16 | 2108 | 65 | ||
| NM_145518.2 | NP_663493.2 | 15 - 16 | 1950 | 65 | ||
| GenBank mRNA | AK034597.1 | - | 15 - 16 | 2019 | 65 | |
| AK036926.1 | - | 15 - 16 | 1815 | 65 | ||
| AK142711.1 | - | 15 - 16 | 1830 | 65 | ||
| AK146814.1 | - | 15 - 16 | 1968 | 65 | ||
| AK167705.1 | - | 15 - 16 | 1798 | 65 | ||
| AK196215.1 | - | 2 - 3 | 197 | 65 | ||
| AK219412.1 | - | 2 - 3 | 197 | 65 | ||
| BC006660.1 | - | 15 - 16 | 1837 | 3501043 | 65 | |
| BC015300.1 | - | 15 - 16 | 1770 | 4236882 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs1
Genomic Map