Assay Details
| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160038.1 | NP_001153510.1 | 7 - 8 | 926 | 62 | |
| NM_001160039.1 | NP_001153511.1 | 7 - 8 | 768 | 62 | ||
| NM_001160040.1 | NP_001153512.1 | 6 - 7 | 822 | 62 | ||
| NM_145518.2 | NP_663493.2 | 6 - 7 | 664 | 62 | ||
| GenBank mRNA | AK034597.1 | - | 6 - 7 | 733 | 62 | |
| AK036926.1 | - | 6 - 7 | 529 | 62 | ||
| AK078590.1 | - | 6 - 7 | 510 | 62 | ||
| AK142711.1 | - | 6 - 7 | 544 | 62 | ||
| AK146814.1 | - | 6 - 7 | 682 | 62 | ||
| AK166604.1 | - | 7 - 8 | 615 | 62 | ||
| AK167705.1 | - | 6 - 7 | 512 | 62 | ||
| AK213867.1 | - | 3 - 4 | 276 | 62 | ||
| BC006660.1 | - | 6 - 7 | 551 | 3501043 | 62 | |
| BC015300.1 | - | 6 - 7 | 484 | 4236882 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs1
Genomic Map