Assay Details
| Gene Symbol: Ica1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029991.1 | - | 1 - 2 | 53 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029991.1 transcripts.
Set Membership: |
|

Ica1l
Genomic Map