Assay Details
| Gene Symbol: Ica1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027407.3 | NP_081683.3 | 3 - 4 | 529 | 112 | |
| GenBank mRNA | AK006525.1 | - | 1 - 2 | 57 | 112 | |
| AK029991.1 | - | 3 - 4 | 301 | 112 | ||
| AK158859.1 | - | 3 - 4 | 365 | 112 | ||
| BC079669.1 | - | 3 - 4 | 529 | 30355976 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006525.1 transcripts.
Set Membership: |
|

Ica1l
Genomic Map