Assay Details
| Gene Symbol: Ica1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027407.3 | NP_081683.3 | 4 - 5 | 662 | 85 | |
| GenBank mRNA | AK029991.1 | - | 4 - 5 | 434 | 85 | |
| AK158859.1 | - | 4 - 5 | 498 | 85 | ||
| BC079669.1 | - | 4 - 5 | 662 | 30355976 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ica1l
Genomic Map