Assay Details
| Gene Symbol: Ica1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027407.3 | NP_081683.3 | 5 - 6 | 863 | 98 | |
| GenBank mRNA | AK006525.1 | - | 3 - 4 | 391 | 98 | |
| AK029991.1 | - | 5 - 6 | 635 | 98 | ||
| AK158859.1 | - | 5 - 6 | 699 | 98 | ||
| BC079669.1 | - | 5 - 6 | 863 | 30355976 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ica1l
Genomic Map