Assay Details
| Gene Symbol: Dera | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172733.1 | NP_766321.1 | 7 - 8 | 779 | 90 | |
| GenBank mRNA | AK012558.1 | - | 7 - 8 | 815 | 90 | |
| AK083298.1 | - | 7 - 8 | 779 | 90 | ||
| AK145820.1 | - | 7 - 8 | 815 | 90 | ||
| AK150273.1 | - | 7 - 8 | 805 | 90 | ||
| AK150712.1 | - | 7 - 8 | 775 | 90 | ||
| AK152462.1 | - | 7 - 8 | 783 | 90 | ||
| AK168204.1 | - | 7 - 8 | 815 | 90 | ||
| AK169418.1 | - | 7 - 8 | 799 | 90 | ||
| AK199271.1 | - | 2 - 3 | 146 | 90 | ||
| BC016218.1 | - | 7 - 8 | 785 | 4484163 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dera
Genomic Map