Assay Details
| Gene Symbol: Ipo8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081113.1 | NP_001074582.1 | 15 - 16 | 1976 | 102 | |
| GenBank mRNA | AK141137.1 | - | 12 - 13 | 3158 | 102 | |
| BC034091.1 | - | 2 - 3 | 268 | 102 | ||
| BC151036.1 | - | 15 - 16 | 1844 | 9087949 | 102 | |
| BC151052.1 | - | 15 - 16 | 1844 | 9087965 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo8
Genomic Map