Assay Details
| Gene Symbol: Arnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037737.2 | NP_001032826.1 | 1 - 2 | 167 | 60 | |
| NM_009709.4 | NP_033839.2 | 1 - 2 | 167 | 60 | ||
| GenBank mRNA | AK028546.1 | - | 1 - 2 | 154 | 60 | |
| AK040475.1 | - | 1 - 2 | 169 | 60 | ||
| AK141362.1 | - | 1 - 2 | 38 | 60 | ||
| AK145415.1 | - | 1 - 2 | 122 | 60 | ||
| AK152473.1 | - | 1 - 2 | 149 | 60 | ||
| AK153355.1 | - | 1 - 2 | 149 | 60 | ||
| BC012870.1 | - | 1 - 2 | 129 | 3964932 | 60 | |
| CJ183661.1 | - | 1 - 2 | 167 | 60 | ||
| U10325.1 | - | 1 - 2 | 31 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028546.1, AK141362.1, AK145415.1, AK152473.1, AK153355.1, BC012870.1, NM_001037737.2, NM_009709.4, U10325.1 transcripts.
Set Membership: |
|

Arnt
Genomic Map