| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800011587.mm.1 |
NM_201354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011587.mm.1 |
NM_201354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011587.mm.1 |
TC1800001603.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800011586.mm.1 |
NM_201354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011586.mm.1 |
NM_201354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011586.mm.1 |
TC1800001603.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800011588.mm.1 |
NM_201354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011588.mm.1 |
NM_201354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011588.mm.1 |
TC1800001603.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800011585.mm.1 |
NM_201354.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011585.mm.1 |
NM_201354.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011585.mm.1 |
TC1800001603.mm.1 |
MTA TC |