Assay Details
| Gene Symbol: Ctif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201354.2 | NP_958742.2 | 9 - 10 | 1777 | 102 | |
| GenBank mRNA | AK172949.1 | - | 2 - 3 | 380 | 102 | |
| BC058104.1 | - | 9 - 10 | 1635 | 6833739 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctif
Genomic Map