Assay Details
| Gene Symbol: Pecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023523.5 | NP_076012.3 | 2 - 3 | 348 | 128 | |
| GenBank mRNA | AF232011.1 | - | 2 - 3 | 254 | 128 | |
| AF242204.1 | - | 2 - 3 | 288 | 128 | ||
| AK010260.1 | - | 2 - 3 | 339 | 128 | ||
| AK135714.1 | - | 2 - 3 | 293 | 128 | ||
| AK162100.1 | - | 2 - 3 | 296 | 128 | ||
| BC013530.1 | - | 2 - 3 | 272 | 4238679 | 128 | |
| BG078542.2 | - | 2 - 3 | 348 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pecr
Genomic Map