Assay Details
| Gene Symbol: Pecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023523.5 | NP_076012.3 | 6 - 7 | 811 | 61 | |
| GenBank mRNA | AF242204.1 | - | 6 - 7 | 751 | 61 | |
| AK010260.1 | - | 6 - 7 | 802 | 61 | ||
| AK090327.1 | - | 6 - 7 | 740 | 61 | ||
| AK162100.1 | - | 6 - 7 | 760 | 61 | ||
| BC013530.1 | - | 6 - 7 | 735 | 4238679 | 61 | |
| BC020329.1 | - | 1 - 2 | 47 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pecr
Genomic Map