Assay Details
| Gene Symbol: Far2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178797.3 | NP_848912.1 | 1 - 2 | 94 | 66 | |
| GenBank mRNA | AK038584.1 | - | 1 - 2 | 47 | 66 | |
| AK043388.1 | - | 1 - 2 | 94 | 66 | ||
| AK043589.1 | - | 1 - 2 | 94 | 66 | ||
| AK137298.1 | - | 1 - 2 | 94 | 66 | ||
| AK161201.1 | - | 1 - 2 | 61 | 66 | ||
| BC055759.1 | - | 1 - 2 | 72 | 6809131 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038584.1, AK043388.1, AK043589.1, AK161201.1, BC055759.1, NM_178797.3 transcripts.
Set Membership: |
|

Far2
Genomic Map