Assay Details
| Gene Symbol: Far2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178797.3 | NP_848912.1 | 4 - 5 | 680 | 83 | |
| GenBank mRNA | AK038584.1 | - | 4 - 5 | 633 | 83 | |
| AK043388.1 | - | 4 - 5 | 680 | 83 | ||
| AK043589.1 | - | 4 - 5 | 680 | 83 | ||
| AK137298.1 | - | 4 - 5 | 680 | 83 | ||
| BC055759.1 | - | 4 - 5 | 658 | 6809131 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Far2
Genomic Map