Assay Details
| Gene Symbol: Adam30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027665.1 | NP_081941.1 | 1 | 199 | 150 | |
| GenBank mRNA | AK077058.1 | - | 352 | 150 | ||
| AY190760.1 | - | 1 | 199 | 150 | ||
| BC113152.1 | - | 1 | 191 | 40059344 | 150 | |
| BC113762.1 | - | 1 | 191 | 40059342 | 150 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Adam30
Genomic Map