Assay Details
| Gene Symbol: Hmgcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008256.4 | NP_032282.2 | 4 - 5 | 908 | 69 | |
| GenBank mRNA | AK004865.1 | - | 4 - 5 | 906 | 69 | |
| AK004902.1 | - | 4 - 5 | 904 | 69 | ||
| AK098104.1 | - | 4 - 5 | 904 | 69 | ||
| BC014714.1 | - | 4 - 5 | 894 | 4216637 | 69 | |
| BC024744.1 | - | 4 - 5 | 892 | 5123821 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcs2
Genomic Map