Assay Details
| Gene Symbol: Hmgcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008256.4 | NP_032282.2 | 5 - 6 | 1069 | 78 | |
| GenBank mRNA | AK004865.1 | - | 5 - 6 | 1067 | 78 | |
| AK004902.1 | - | 5 - 6 | 1065 | 78 | ||
| AK098104.1 | - | 5 - 6 | 1065 | 78 | ||
| BC014714.1 | - | 5 - 6 | 1055 | 4216637 | 78 | |
| BC024744.1 | - | 5 - 6 | 1053 | 5123821 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcs2
Genomic Map