Assay Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007505.2 | NP_031531.1 | 3 - 4 | 593 | 67 | |
| GenBank mRNA | AK043976.1 | - | 3 - 4 | 361 | 67 | |
| AK076572.1 | - | 3 - 4 | 361 | 67 | ||
| AK098155.1 | - | 3 - 4 | 371 | 67 | ||
| AK146797.1 | - | 3 - 4 | 364 | 67 | ||
| AK150426.1 | - | 3 - 4 | 364 | 67 | ||
| AK150843.1 | - | 3 - 4 | 361 | 67 | ||
| AK151004.1 | - | 3 - 4 | 361 | 67 | ||
| AK151128.1 | - | 3 - 4 | 370 | 67 | ||
| AK151224.1 | - | 3 - 4 | 368 | 67 | ||
| AK151920.1 | - | 3 - 4 | 365 | 67 | ||
| AK152054.1 | - | 3 - 4 | 372 | 67 | ||
| AK152890.1 | - | 3 - 4 | 364 | 67 | ||
| AK153167.1 | - | 3 - 4 | 365 | 67 | ||
| AK153282.1 | - | 3 - 4 | 361 | 67 | ||
| AK157529.1 | - | 1 - 2 | 142 | 67 | ||
| AK159491.1 | - | 3 - 4 | 361 | 67 | ||
| AK159540.1 | - | 3 - 4 | 361 | 67 | ||
| AK159758.1 | - | 3 - 4 | 364 | 67 | ||
| AK160043.1 | - | 3 - 4 | 371 | 67 | ||
| AK164110.1 | - | 3 - 4 | 364 | 67 | ||
| AK164193.1 | - | 1 - 2 | 67 | 67 | ||
| AK166709.1 | - | 3 - 4 | 364 | 67 | ||
| AK166812.1 | - | 3 - 4 | 377 | 67 | ||
| AK167159.1 | - | 3 - 4 | 361 | 67 | ||
| AK167863.1 | - | 3 - 4 | 341 | 67 | ||
| AK168198.1 | - | 3 - 4 | 371 | 67 | ||
| AK168369.1 | - | 3 - 4 | 361 | 67 | ||
| AK168617.1 | - | 3 - 4 | 361 | 67 | ||
| AK168879.1 | - | 3 - 4 | 361 | 67 | ||
| AK168890.1 | - | 3 - 4 | 361 | 67 | ||
| AK168932.1 | - | 3 - 4 | 361 | 67 | ||
| AK169080.1 | - | 3 - 4 | 364 | 67 | ||
| AK169084.1 | - | 3 - 4 | 364 | 67 | ||
| AK169105.1 | - | 3 - 4 | 361 | 67 | ||
| AK169142.1 | - | 3 - 4 | 406 | 67 | ||
| AK169300.1 | - | 3 - 4 | 361 | 67 | ||
| AK169308.1 | - | 3 - 4 | 366 | 67 | ||
| AK169414.1 | - | 3 - 4 | 364 | 67 | ||
| AK196974.1 | - | 1 - 2 | 120 | 67 | ||
| BC014854.1 | - | 3 - 4 | 348 | 4500893 | 67 | |
| BC055892.1 | - | 3 - 4 | 798 | 67 | ||
| L01062.1 | - | 3 - 4 | 348 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5a1
Genomic Map