Assay Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007505.2 | NP_031531.1 | 4 - 5 | 766 | 57 | |
| GenBank mRNA | AK043976.1 | - | 4 - 5 | 534 | 57 | |
| AK076572.1 | - | 4 - 5 | 534 | 57 | ||
| AK098155.1 | - | 4 - 5 | 544 | 57 | ||
| AK146797.1 | - | 4 - 5 | 537 | 57 | ||
| AK150426.1 | - | 4 - 5 | 537 | 57 | ||
| AK150843.1 | - | 4 - 5 | 534 | 57 | ||
| AK151004.1 | - | 4 - 5 | 534 | 57 | ||
| AK151128.1 | - | 4 - 5 | 543 | 57 | ||
| AK151224.1 | - | 4 - 5 | 541 | 57 | ||
| AK151920.1 | - | 4 - 5 | 538 | 57 | ||
| AK152054.1 | - | 4 - 5 | 545 | 57 | ||
| AK152890.1 | - | 4 - 5 | 537 | 57 | ||
| AK153167.1 | - | 4 - 5 | 538 | 57 | ||
| AK153282.1 | - | 4 - 5 | 534 | 57 | ||
| AK157529.1 | - | 2 - 3 | 315 | 57 | ||
| AK159491.1 | - | 4 - 5 | 534 | 57 | ||
| AK159540.1 | - | 4 - 5 | 534 | 57 | ||
| AK159758.1 | - | 4 - 5 | 537 | 57 | ||
| AK160043.1 | - | 4 - 5 | 544 | 57 | ||
| AK164110.1 | - | 4 - 5 | 537 | 57 | ||
| AK164193.1 | - | 2 - 3 | 240 | 57 | ||
| AK166709.1 | - | 4 - 5 | 537 | 57 | ||
| AK166812.1 | - | 4 - 5 | 550 | 57 | ||
| AK167159.1 | - | 4 - 5 | 534 | 57 | ||
| AK167863.1 | - | 4 - 5 | 514 | 57 | ||
| AK168198.1 | - | 4 - 5 | 544 | 57 | ||
| AK168369.1 | - | 4 - 5 | 534 | 57 | ||
| AK168617.1 | - | 4 - 5 | 534 | 57 | ||
| AK168879.1 | - | 4 - 5 | 534 | 57 | ||
| AK168890.1 | - | 4 - 5 | 534 | 57 | ||
| AK168932.1 | - | 4 - 5 | 534 | 57 | ||
| AK169080.1 | - | 4 - 5 | 537 | 57 | ||
| AK169084.1 | - | 4 - 5 | 537 | 57 | ||
| AK169105.1 | - | 4 - 5 | 534 | 57 | ||
| AK169142.1 | - | 4 - 5 | 579 | 57 | ||
| AK169300.1 | - | 4 - 5 | 534 | 57 | ||
| AK169308.1 | - | 4 - 5 | 539 | 57 | ||
| AK169414.1 | - | 4 - 5 | 537 | 57 | ||
| BC014854.1 | - | 4 - 5 | 521 | 4500893 | 57 | |
| BC020417.1 | - | 4 - 5 | 521 | 57 | ||
| BC055892.1 | - | 4 - 5 | 971 | 57 | ||
| L01062.1 | - | 4 - 5 | 521 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5a1
Genomic Map