Assay Details
| Gene Symbol: Haus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146089.2 | NP_666201.1 | 4 - 5 | 505 | 62 | |
| GenBank mRNA | AK146923.1 | - | 4 - 5 | 502 | 62 | |
| BC024400.1 | - | 4 - 5 | 505 | 5026575 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Haus1
Genomic Map