Assay Details
| Gene Symbol: Haus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146089.2 | NP_666201.1 | 7 - 8 | 771 | 94 | |
| GenBank mRNA | AK076912.1 | - | 7 - 8 | 751 | 94 | |
| AK146923.1 | - | 7 - 8 | 768 | 94 | ||
| BC024400.1 | - | 7 - 8 | 771 | 5026575 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849646 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849647 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849645 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849648 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849679 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170696 | AK146923.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170696 | AK147599.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170696 | BC024400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170696 | NM_146089.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170696 | AK146923.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170696 | AK147599.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170696 | BC024400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170696 | NM_146089.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116222 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165029 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170946 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170696 | AK146923.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170696 | AK147599.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170696 | BC024400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170696 | NM_146089.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170696 | AK146923.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170696 | AK147599.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170696 | BC024400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P170696 | NM_146089.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170696 | AK146923.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170696 | AK147599.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170696 | BC024400.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170696 | NM_146089.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170696 | AK146923.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170696 | AK147599.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170696 | BC024400.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170696 | NM_146089.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31849648 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09060170 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170946 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165029 | - | - |
| Whole Mouse Genome G4122A | A_51_P170696 | AK146923.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170696 | AK147599.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170696 | BC024400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170696 | NM_146089.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170696 | AK146923.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170696 | AK147599.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170696 | BC024400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P170696 | NM_146089.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139192 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128685 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170946 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165029 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13903837 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13915886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13903828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13903827 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13845994 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849648 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849646 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849645 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849679 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31849647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31849705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13848099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13693974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13903838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13834942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14122620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13736844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13664280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13903837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14006467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14093968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31849624 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10459847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459853 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10459852 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424955_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10459844 | - | - |
| Mouse Genome 430 2.0 Array | 1424955_at | - | - |
| Mouse Genome 430 2.0 Array | 1424955_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225745_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225745_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800011728.mm.1 | NM_146089.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800011728.mm.1 | NM_146089.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800011728.mm.1 | TC1800001632.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800011729.mm.1 | NM_146089.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800011729.mm.1 | NM_146089.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800011729.mm.1 | TC1800001632.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424955_PM_at | - | - |
| Mouse Expression Set 430 | 1424955_at | - | - |
| Murine Genome U74v2 Set | 137986_at | - | - |
| Murine Genome U74v2 Set | 137986_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424955_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424955_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424955_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424955_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 916971 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2480040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2480040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002480040 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250852 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250852 | - | - |
| mouse-6 v1.1 (Illumina) | scl51245.13.1_81-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2480040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51245.13.1_81-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1250852 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250852 | - | - |

Haus1
Genomic Map