Assay Details
| Gene Symbol: Fbxo38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134136.3 | NP_598897.2 | 8 - 9 | 1128 | 60 | |
| GenBank mRNA | AK031096.1 | - | 8 - 9 | 1078 | 60 | |
| AK031347.1 | - | 8 - 9 | 1059 | 60 | ||
| AK034761.1 | - | 3 - 4 | 312 | 60 | ||
| AK089046.1 | - | 8 - 9 | 1100 | 60 | ||
| AY267463.1 | - | 8 - 9 | 1061 | 60 | ||
| BC056348.1 | - | 8 - 9 | 1080 | 5721407 | 60 | |
More Information
Set Membership: |
|

Fbxo38
Genomic Map