Assay Details
| Gene Symbol: Nfib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113209.2 | NP_001106680.1 | 5 - 6 | 1829 | 84 | |
| NM_001113210.2 | NP_001106681.1 | 5 - 6 | 1829 | 84 | ||
| NM_001286127.1 | NP_001273056.1 | 5 - 6 | 1829 | 84 | ||
| NM_001286131.1 | NP_001273060.1 | 5 - 6 | 1829 | 84 | ||
| NM_008687.6 | NP_032713.3 | 5 - 6 | 1829 | 84 | ||
| GenBank mRNA | AK034261.1 | - | 5 - 6 | 1361 | 84 | |
| AK083167.1 | - | 5 - 6 | 1174 | 84 | ||
| AK084941.1 | - | 5 - 6 | 1434 | 84 | ||
| AK087480.1 | - | 5 - 6 | 1361 | 84 | ||
| AK145267.1 | - | 4 - 5 | 535 | 84 | ||
| AK146480.1 | - | 5 - 6 | 1346 | 84 | ||
| AY035852.1 | - | 5 - 6 | 834 | 84 | ||
| AY035853.1 | - | 5 - 6 | 834 | 84 | ||
| BC014290.1 | - | 5 - 6 | 866 | 4038233 | 84 | |
| BC023366.1 | - | 5 - 6 | 966 | 84 | ||
| BC062908.1 | - | 5 - 6 | 1151 | 6815775 | 84 | |
| BE862566.1 | - | 4 - 5 | 480 | 84 | ||
| U57634.1 | - | 5 - 6 | 1035 | 84 | ||
| Y07685.1 | - | 5 - 6 | 1761 | 84 | ||
| Y07686.1 | - | 5 - 6 | 1341 | 84 | ||
| Y07687.1 | - | 5 - 6 | 1822 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfib
Genomic Map