| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400023411.mm.1 |
NM_001017427.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400023411.mm.1 |
NM_001017427.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400023411.mm.1 |
TC0400002862.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400023412.mm.1 |
NM_001017427.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400023412.mm.1 |
NM_001017427.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400023412.mm.1 |
TC0400002862.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400023413.mm.1 |
NM_001017427.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400023413.mm.1 |
NM_001017427.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400023413.mm.1 |
TC0400002862.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400023414.mm.1 |
NM_001017427.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400023414.mm.1 |
NM_001017427.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400023414.mm.1 |
TC0400002862.mm.1 |
MTA TC |