Assay Details
| Gene Symbol: Stk32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178749.3 | NP_848864.1 | 5 - 6 | 676 | 95 | |
| GenBank mRNA | AK036266.1 | - | 5 - 6 | 677 | 95 | |
| AK042599.1 | - | 5 - 6 | 790 | 95 | ||
| AK044474.1 | - | 5 - 6 | 686 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk32a
Genomic Map