Assay Details
| Gene Symbol: Stk32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178749.3 | NP_848864.1 | 8 - 9 | 901 | 61 | |
| GenBank mRNA | AK036266.1 | - | 8 - 9 | 902 | 61 | |
| AK042599.1 | - | 8 - 9 | 1015 | 61 | ||
| AK044474.1 | - | 8 - 9 | 911 | 61 | ||
| BC055002.1 | - | 831 | 6313304 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk32a
Genomic Map