Assay Details
| Gene Symbol: Gnal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010307.3 | NP_034437.1 | 6 - 7 | 806 | 98 | |
| NM_177137.5 | NP_796111.2 | 6 - 7 | 971 | 98 | ||
| GenBank mRNA | AK158831.1 | - | 6 - 7 | 807 | 98 | |
| AY179169.1 | - | 6 - 7 | 540 | 98 | ||
| BC078439.1 | - | 6 - 7 | 933 | 5685967 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnal
Genomic Map