Assay Details
| Gene Symbol: Tcerg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039474.1 | NP_001034563.1 | 7 - 8 | 1443 | 90 | |
| NM_001289526.1 | NP_001276455.1 | 6 - 7 | 1380 | 90 | ||
| NR_110344.1 | 7 - 8 | 1443 | 90 | |||
| GenBank mRNA | AB023485.1 | - | 7 - 8 | 1407 | 90 | |
| AK010571.1 | - | 6 - 7 | 1362 | 90 | ||
| AK013830.1 | - | 6 - 7 | 1365 | 90 | ||
| AK076993.1 | - | 7 - 8 | 1422 | 90 | ||
| AK150312.1 | - | 6 - 7 | 1370 | 90 | ||
| AK168434.1 | - | 7 - 8 | 1421 | 90 | ||
| BC039185.1 | - | 6 - 7 | 1333 | 3982736 | 90 | |
| BC040284.1 | - | 6 - 7 | 1343 | 4460736 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcerg1
Genomic Map