Assay Details
| Gene Symbol: Fgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010197.3 | NP_034327.1 | 3 - 4 | 457 | 69 | |
| GenBank mRNA | AK035330.1 | - | 3 - 4 | 458 | 69 | |
| AK142163.1 | - | 4 - 5 | 534 | 69 | ||
| BC027001.1 | - | 3 - 4 | 446 | 69 | ||
| BC037601.1 | - | 3 - 4 | 476 | 5137246 | 69 | |
| M30641.1 | - | 2 - 3 | 270 | 69 | ||
| U67610.1 | - | 3 - 4 | 423 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf1
Genomic Map